Learn More
BRAND™ LDPE Wide Neck Wash Bottle
Catalog Number 15517831
Click to view available options
Color:
Orange
White
Yellow
High-quality wash bottles with integrated wash stream and sealing cone closure
  • Flexible
  • Wide mouth
  • Large opening for convenient filling
  • Label with designation of contents and general safety statements in English (with CAS numbers and NFPA codes) and German (with CAS numbers)
  • Capacity (Metric) 250 mL
    Color Yellow
    Material LDPE
    Quantity 5