Learn More
Lead(II) Nitrate, Supelco™
Product Code. 15817580
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Product Code. Quantity Unit Size
15817580 1 kg 1kg
1 options
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
Product Code. 15817580 Supplier Supelco Supplier No. 1073981000
 Restricted Item
Beräknad leverans: 26-11-2026
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
For analysis EMSURE™ ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 458°C to 459°C
CAS 10099-74-8
Quantity 1 kg
Molecular Formula Pb(NO3)2
MDL Number MFCD00011153
Synonym Lead(II) nitrate, Lead(II) nitrate
Molecular Weight (g/mol) 331.21
Grade Research
Density 4.49 g/cm3 at 20°C
Chemical Name or Material Lead(II) Nitrate