Learn More
Lead(II) Oxide, Extra Pure, MilliporeSigma™
Product Code. 15864640
Change view
Click to view available options
Quantity:
5 kg
50 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
50kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15864640 50 kg Plastic drum 50kg
15854640 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15864640 Supplier Supelco Supplier No. 1056589050
 Restricted Item
Beräknad leverans: 26-11-2026
Chemical Identifiers
CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
IUPAC Name plumbanone
SMILES O=[Pb]
Specifications
Physical Form Solid
Quantity 50 kg
Odor Odorless
Packaging Plastic drum
Appearance Yellow solid
UN Number UN 2291
Formula Weight 223.19
Percent Purity ≥99.0%
Grade Extra Pure
Chemical Name or Material Lead(II) oxide