Läs mer
Tocris Bioscience™ Lenalidomide 4'-PEG1-azide
Beskrivning
Lenalidomide 4'-PEG1-azide is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG1 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3-[4-[[2-(2-Azidoethoxy)ethyl]amino]-1,3-dihydro-1-oxo-2 H-isoindol-2-yl]-2,6-piperidinedione |
| CAS | 2399455-45-7 |
| Kvantitet | 25 mg |
| Mål | Degrader Building Blocks |
| Molekylformel | C17H20N6O4 |
| Renhet | 0.95 |