Läs mer
3M™ Lens Cover Peel-Offs 7992
Handla allt 3M Produkter
:
Color: Transparent
Beskrivning
- Transparent Peel-Off lens cover for the 3M™ Full Face Mask 7907S or 7900PF
- Self-adhesive fastening allows for easy application and removal
- Protects the visor from contaminants and scratches
- For use with 3M™ Powered Air Respirator 7900PF
- Quick and easy to fit
- Increase the effective service life of your equipment
Specifikationer
Specifikationer
| Certifikat och efterlevnad | CE Approved |
| Färg | Transparent |
| Produkttyp | Replacement Lens Cover |