Läs mer
Lenvatinib Mesylate, TRC
Produktkod. 30480240
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480240 25 mg
30482174 50 mg
30477975 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30480240 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL21598025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 857890-39-2
Molekylformel C21 H19 Cl N4 O4 . C H4 O3 S
Molekylvikt (g/mol) 522.959
Synonym 4-[3-Chloro-4-[(cyclopropylaminocarbonyl)amino]phenoxy]-7-methoxy-6-quinolinecarboxamide Mesylate, E 7080 Mesylate, Kisplyx, Lenvatinib Mesylate, Lenvatinib Methanesulfonate, Lenvima
IUPAC-namn 4-[3-chloro-4-(cyclopropylcarbamoylamino)phenoxy]-7-methoxyquinoline-6-carboxamide;methanesulfonic acid
LEDER COc1cc2nccc(Oc3ccc(NC(=O)NC4CC4)c(Cl)c3)c2cc1C(=O)N.CS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Lenvatinib Mesylate
Kvantitet 25 mg
InChI formel InChI=1S/C21H19ClN4O4.CH4O3S/c1-29-19-10-17-13(9-14(19)20(23)27)18(6-7-24-17)30-12-4-5-16(15(22)8-12)26-21(28)25-11-2-3-11;1-5(2,3)4/h4-11H,2-3H2,1H3,(H2,23,27)(H2,25,26,28);1H3,(H,2,3,4)
Formel vikt 522.098