Läs mer
Lenvatinib N-Oxide, TRC
Produktkod. 30472773
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30472773 5 mg
1 options
Produktkod. 30472773 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3295005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lenvatinib N-Oxide
CAS 1788901-86-9
Molekylformel C21H19ClN4O5
Kvantitet 5 mg
Synonym 4-[3-Chloro-4-[[(cyclopropylamino)carbonyl]amino]phenoxy]-7-methoxy-6-quinolinecarboxamide 1-Oxide, Lenvatinib Metabolite M3
InChI formel InChI=1S/C21H19ClN4O5/c1-30-19-10-17-13(9-14(19)20(23)27)18(6-7-26(17)29)31-12-4-5-16(15(22)8-12)25-21(28)24-11-2-3-11/h4-11H,2-3H2,1H3,(H2,23,27)(H2,24,25,28)
LEDER COc1cc2c(cc1C(=O)N)c(Oc3ccc(NC(=O)NC4CC4)c(Cl)c3)cc[n+]2[O-]
Molekylvikt (g/mol) 442.85