Läs mer
Leonurine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Leonurine hydrochloride |
| CAS | 24735-18-0 |
| Molekylformel | C14 H21 N3 O5 . Cl H |
| Kvantitet | 250 mg |
| Synonym | Benzoic acid, 4-hydroxy-3,5-dimethoxy-, 4-[(aminoiminomethyl)amino]butyl ester, hydrochloride (1:1), Benzoic acid, 4-hydroxy-3,5-dimethoxy-, 4-[(aminoiminomethyl)amino]butyl ester, monohydrochloride (9CI), Benzoic acid, 4-hydroxy-3,5-dimethoxy-, ester with (4-hydroxybutyl)guanidine monohydrochloride (8CI), Leonurine, hydrochloride (7CI), Guanidine, (4-hydroxybutyl)-, 4-hydroxy-3,5-dimethoxybenzoate (ester), monohydrochloride (8CI) |
| InChI formel | InChI=1S/C14H21N3O5.ClH/c1-20-10-7-9(8-11(21-2)12(10)18)13(19)22-6-4-3-5-17-14(15)16;/h7-8,18H,3-6H2,1-2H3,(H4,15,16,17);1H |
| LEDER | Cl.COc1cc(cc(OC)c1O)C(=O)OCCCCNC(=N)N |
| IUPAC-namn | 4-carbamimidamidobutyl 4-hydroxy-3,5-dimethoxybenzoate;hydrochloride |
| Molekylvikt (g/mol) | 347.79 |
| Formel vikt | 347.1248 |