Läs mer
Leonurine, TRC
Produktkod. 30475209
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30475209 1 g
1 options
Produktkod. 30475209 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3296031G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Leonurine
CAS 24697-74-3
Molekylformel C14H21N3O5
Kvantitet 1 g
Synonym 4-Guanidino-1-butanol Syringate, 4-Hydroxy-3,5-dimethoxybenzoic Acid, Δ-guanidinobutyl Ester, Leonurin, SCM 198, Syringic Acid Δ-guanidinobutyl Ester, [4-(4-Hydroxy-3,5-dimethoxybenzoyloxy)butyl]guanidine, 4-Guanidinobutyl 4-hydroxy-3,5-dimethoxybenzoate
InChI formel InChI=1S/C14H21N3O5/c1-20-10-7-9(8-11(21-2)12(10)18)13(19)22-6-4-3-5-17-14(15)16/h7-8,18H,3-6H2,1-2H3,(H4,15,16,17)
LEDER CN(C)CCc1c2c3c4c(c(OC)ccc4C(=O)Oc3c(OC)c1)OC2=O
Molekylvikt (g/mol) 311.33