Learn More
Abnova™ LEPRE1 Recombinant Protein
Human LEPRE1 full-length ORF (AAH15309, 1 a.a. - 390 a.a.) recombinant protein with GST-tag at N-terminal
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH15309 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 64175 |
Molecular Weight (g/mol) | 68.53 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16173463
|
Abnova™
H00064175-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16183463
|
Abnova™
H00064175-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes an enzyme that is a member of the collagen prolyl hydroxylase family. These enzymes are localized to the endoplasmic reticulum and their activity is required for proper collagen synthesis and assembly.
- Molecular weight: 68.53kDa
- Preparation method: in vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8 in the elution buffer
- Quality Control Testing: 12.5% SDS-PAGE stained with Coomassie Blue
Sequence:
MLGEEHTRSIGPRESAKEYRQRSLLEKELLFFAYDVFGIPFVDPDSWTPEEVIPKRLQEKQKSERETAVRISQEIGNLMKEIETLVEEKTKESLDVSRLTREGGPLLYEGISLTMNSKLLNGSQRVVMDGVISDHECQELQRLTNVAATSGDGYRGQTSPHTPNEKFYGVTVFKALKLGQEGKVPLQSAHLYYNVTEKVRRIMESYFRLDTPLYFSYSHLVCRTAIEEVQAERKDDSHPVHVDNCILNAETLVCVKEPPAYTFRDYSAILYLNGDFDGGNFYFTELDAKTVTAEVQPQCGRAVGFSSGTENPHGVKAVTRGQRCAIALWFTLDPRHSERDRVQADDLVKMLFSPEEMDLSQEQPLDAQQGPPEPAQESLSGSESKPKDEL
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH15309 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
68.53 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LEPRE1 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
64175 | |
LEPRE1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLGEEHTRSIGPRESAKEYRQRSLLEKELLFFAYDVFGIPFVDPDSWTPEEVIPKRLQEKQKSERETAVRISQEIGNLMKEIETLVEEKTKESLDVSRLTREGGPLLYEGISLTMNSKLLNGSQRVVMDGVISDHECQELQRLTNVAATSGDGYRGQTSPHTPNEKFYGVTVFKALKLGQEGKVPLQSAHLYYNVTEKVRRIMESYFRLDTPLYFSYSHLVCRTAIEEVQAERKDDSHPVHVDNCILNAETLVCVKEPPAYTFRDYSAILYLNGDFDGGNFYFTELDAKTVTAEVQPQCGRAVGFSSGTENPHGVKAVTRGQRCAIALWFTLDPRHSERDRVQADDLVKMLFSPEEMDLSQEQPLDAQQGPPEPAQESLSGSESKPKDEL | |
GROS1/MGC117314/P3H1 | |
LEPRE1 | |
Wheat Germ (in vitro) | |
GST |