Läs mer
Lerociclib dihydrochloride, MedChemExpress
Produktkod. 16474608
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
25mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16474608 10 mg -
16470907 50 mg -
16445456 100 mg -
16427327 5 mg -
16403166 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16474608 Leverantör Medchem Express Leverantörsnummer HY112272A
MedChemExpress Lerociclib dihydrochloride (G1T38 dihydrochloride) is a potent and selective inhibitor of CDK4/CDK6, with IC50s of 1 nM and 2 nM for CDK4/CyclinD1 and CDK6/CyclinD3, respectively.
Lerociclib dihydrochloride (G1T38 dihydrochloride) is a potent and selective inhibitor of CDK4/CDK6, with IC50s of 1 nM and 2 nM for CDK4/CyclinD1 and CDK6/CyclinD3, respectively.
Specifikationer
Kemiskt namn eller material Lerociclib dihydrochloride
CAS 2097938-59-3
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C26H36Cl2N8O
Kvantitet 10 mg
Synonym G1T38 dihydrochloride
Löslighetsinformation H2O : 4 mg/mL (7.31 mM; Need ultrasonic)
LEDER O=C1NCC2(N3C1=CC4=CN=C(NC5=NC=C(N6CCN(C(C)C)CC6)C=C5)N=C43)CCCCC2.Cl.Cl
Molekylvikt (g/mol) 547.52
Formel vikt 547.52
Procent renhet 99.74%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease