Läs mer
Letrozole Diacid Impurity, TRC
Produktkod. 30477304
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477304 10 mg
30483453 100 mg
30468523 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30477304 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL33022010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1644566-39-1
Molekylformel C17H13N3O4
Molekylvikt (g/mol) 323.3
Synonym Benzoic acid, 4,4'-(1H-1,2,4-triazol-1-ylmethylene)bis-, 4,4'-(1H-1,2,4-Triazol-1-ylmethylene)bis[benzoic acid], 4,4'-(1H-1,2,4-Triazol-1-ylmethylene)dibenzoic Acid, 4,4'-(1H-1,2,4-triazol-1-ylmethylene)bis-benzoic Acid
IUPAC-namn 4-[(4-carboxyphenyl)-(1,2,4-triazol-1-yl)methyl]benzoic acid
LEDER OC(=O)c1ccc(cc1)C(c2ccc(cc2)C(=O)O)n3cncn3
Specifikationer
Kemiskt namn eller material Letrozole Diacid Impurity
Kvantitet 10 mg
InChI formel InChI=1S/C17H13N3O4/c21-16(22)13-5-1-11(2-6-13)15(20-10-18-9-19-20)12-3-7-14(8-4-12)17(23)24/h1-10,15H,(H,21,22)(H,23,24)
Formel vikt 323.0906