Läs mer
Thermo Scientific Chemicals [Leu13] Motilin, Human, Porcine
Produktkod. 15488539
Kvantitet:
0.5 mg
1 mg
Förpackningsstorlek:
0.5mg
1mg
Produktkod. 15488539
Brand: Thermo Scientific Chemicals J66962.M^
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
For cellular and molecular biology applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifikationer
Art Human, Porcine
Reningsmetod HPLC
Innehåll och lagring -30°C to -10°C
Formatera Solid
Molekylvikt (g/mol) 2681.10
Produkttyp [Leu13] Motilin, human, porcine
Kvantitet 1 mg
Regulatorisk status RUO - research use only
Sekvens Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Leu-Gln-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln