Läs mer
Novus Biologicals™ Leucine Aminopeptidase (LAP) Activity Assay Kit (Fluorometric)
Produktkod. 18291262 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18291262 100 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 18291262 Leverantör Novus Biologicals™ Leverantörsnummer NBP259745
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This kit provides a quick, sensitive and easy way for measuring total LAP activity in various samples.
In this assay, LAPs hydrolyze leucine from the fluorescent probe and the amount of fluorescent probe detected is used to determine the total activity of the LAP enzymes. Detection method: Fluorescence (Ex/Em = 368/460 nm). The assay is simple to perform, high-throughput adaptable and can detect less than 0.1 mU of LAP activity.
Kit Contents:
  • AMC-Standard (1 mM)
  • LAP Assay Buffer
  • LAP Positive Control
  • LAP Substrate
TRUSTED_SUSTAINABILITY
Specifikationer
Innehåll och lagring Store at -20°C in the dark. Avoid freeze-thaw cycles.
Kvantitet 100 Assays
Mål Leucine Aminopeptidase (LAP) Activity