Läs mer
Leuprolide Acetate EP Impurity H, TRC
Produktkod. 30138291
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138291 10 mg
30004756 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138291 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL33063510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 112710-58-4
Synonym Luteinizing hormone-releasing factor (swine), 6-D-leucine-7-D-leucine-9-(N-ethyl-L-prolinamide)-10-deglycinamide- (9CI)∣Luteinizing hormone-releasing factor (pig), 6-D-leucine-7-D-leucine-9-(N-ethyl-L-prolinamide)-10-deglycinamide-∣Leuprolide Acetate EP Impurity H
Specifikationer
Formatera Neat
Molekylformel C61H85F3N16O14
MDL-nummer MFCD18782255
Molekylvikt (g/mol) 1209.4, 1323.44
InChI-nyckel VIKVLAJTEHLRFY-DYEBRENCSA-N
LEDER OC(=O)C(F)(F)F.CCNC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCN=C(N)N)NC(=O)[C@@H](CC(C)C)NC(=O)[C@@H](CC(C)C)NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)[C@H](CO)NC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)[C@H](CC1=CN=CN1)NC(=O)[C@@H]1CCC(=O)N1
IUPAC-namn (2S)-1-[(2S)-5-[(diaminomethylidene)amino]-2-[(2R)-2-[(2R)-2-[(2S)-2-[(2S)-3-hydroxy-2-[(2S)-2-[(2S)-3-(1H-imidazol-5-yl)-2-{[(2S)-5-oxopyrrolidin-2-yl]formamido}propanamido]-3-(1H-indol-3-yl)propanamido]propanamido]-3-(4-hydroxyphenyl)propanamido]-4-methylpentanamido]-4-methylpentanamido]pentanoyl]-N-ethylpyrrolidine-2-carboxamide; trifluoroacetic acid
Produkttyp Impurity