Läs mer
Leuprolide Acetate, TRC
Produktkod. 30136798
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136798 25 mg
30008607 10 mg
30009201 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136798 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL33060025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 88793-81-1
Synonym 6-D-Leucine-9-(N-ethyl-L-prolinamide)-10-deglycinamideluteinizing Hormone-Releasing Factor Acetate(Pig)∣A 43818∣Abbott 43818∣Leuprorelin Acetate∣Lupron∣Procren Depot∣Procrin∣Prostap∣TAP 144∣TAP 144SR
Specifikationer
Renhet >95% (HPLC)
Formatera Neat
Molekylvikt (g/mol) 1269.4
Produkttyp Metabolite∣API