Läs mer
Leurosine Sulfate, TRC
Produktkod. 30476777
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30476777 1 mg
1 options
Produktkod. 30476777 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3308001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Leurosine Sulfate
CAS 54081-68-4
Molekylformel 2 C46 H56 N4 O9 . H2 O4 S
Kvantitet 1 mg
Synonym Vincaleukoblastine, 4'-deoxy-3',4'-epoxy-, (3'α,4'α)-, sulfate (2:1) (ACI), 1H-Indolizino[8,1-cd]carbazole, vincaleukoblastine deriv. (ZCI), 2H-3,13-Methanooxireno[9,10]azacycloundecino[5,4-b]indole, vincaleukoblastine deriv. (ZCI), Vincaleukoblastine, 4'-deoxy-3',4'-epoxy-, (3'α,4'α)-, sulfate (2:1) (salt) (9CI), Leurosine sulfate, Lilly 32645, Vinleurosine sulfate, (3'α,4'α)-4'-Deoxy-3',4'-epoxyvincaleukoblastine hemisulfate, Leurosine hemisulfate, Vinleurosine hemisulfate
InChI formel InChI=1S/2C46H56N4O9.H2O4S/c2*1-8-42-16-12-18-50-20-17-44(37(42)50)30-21-31(34(55-5)22-33(30)48(4)38(44)46(54,41(53)57-7)39(42)58-26(3)51)45(40(52)56-6)23-27-24-49(25-43(9-2)36(27)59-43)19-15-29-28-13-10-11-14-32(28)47-35(29)45;1-5(2,3)4/h2*10-14,16,21-22,27,36-39,47,54H,8-9,15,17-20,23-25H2,1-7H3;(H2,1,2,3,4)/t2*27-,36-,37-,38+,39+,42+,43+,44+,45-,46-;/m00./s1
LEDER CC[C@@]12CN3CCc4c([nH]c5ccccc45)[C@@](C[C@@H](C3)[C@@H]1O2)(C(=O)OC)c6cc7c(cc6OC)N(C)[C@H]8[C@](O)([C@H](OC(=O)C)[C@]9(CC)C=CCN%10CC[C@]78[C@H]9%10)C(=O)OC.CC[C@@]%11%12CN%13CCc%14c([nH]c%15ccccc%14%15)[C@@](C[C@@H](C%13)[C@@H]%11O%12)(C(=O)OC)c%16cc%17c(cc%16OC)N(C)[C@H]%18[C@](O)([C@H](OC(=O)C)[C@]%19(CC)C=CCN%20CC[C@]%17%18[C@H]%19%20)C(=O)OC.OS(=O)(=O)O
IUPAC-namn methyl (13S,15S,16R,18S)-13-[(1R,9R,10S,11R,12R,19R)-11-acetyloxy-12-ethyl-10-hydroxy-5-methoxy-10-methoxycarbonyl-8-methyl-8,16-diazapentacyclo[10.6.1.01,9.02,7.016,19]nonadeca-2,4,6,13-tetraen-4-yl]-18-ethyl-17-oxa-1,11-diazapentacyclo[13.4.1.04,12.05,10.016,18]icosa-4(12),5(10),6,8-tetraene-13-carboxylate;sulfuric acid
Molekylvikt (g/mol) 1716
Formel vikt 1714.7768