Läs mer
Levamlodipine Besylate, TRC
Produktkod. 30472661
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30472661 250 mg
1 options
Produktkod. 30472661 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL246725250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Levamlodipine Besilate
CAS 150566-71-5
Molekylformel C20 H25 Cl N2 O5 . C6 H6 O3 S
Kvantitet 250 mg
Synonym Levamlodipine Besylate, 3,5-Pyridinedicarboxylic acid, 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-, 3-ethyl 5-methyl ester, (4S)-, benzenesulfonate (1:1), 3,5-Pyridinedicarboxylic acid, 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-, 3-
InChI formel InChI=1S/C20H25ClN2O5.C6H6O3S/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21;7-10(8,9)6-4-2-1-3-5-6/h5-8,17,23H,4,9-11,22H2,1-3H3;1-5H,(H,7,8,9)/t17-;/m0./s1
LEDER CCOC(=O)C1=C(COCCN)NC(=C([C@@H]1c2ccccc2Cl)C(=O)OC)C.OS(=O)(=O)c3ccccc3
IUPAC-namn benzenesulfonic acid;3-O-ethyl 5-O-methyl (4S)-2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvikt (g/mol) 567.051
Formel vikt 566.149