Läs mer
Tocris Bioscience™ Levcromakalim
GREENER_CHOICE
Produktkod. 18063912
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18063912 50 mg
18005402 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18063912 Leverantör Tocris Bioscience™ Leverantörsnummer 1378/50
Kir6 (KATP) channel opener; active enantiomer of cromakalim (Cat. No. 1377)
Levcromakalim is an active enantiomer of the prototypical Kir6 (KATP) channel opener Cromakalim. Hypotensive and airways relaxant. IC50 = 490 nM in guinea pig trachea.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 94535-50-9
Molekylformel C16H18N2O3
Specifikationer
Kemiskt namn eller material Levcromakalim
Kvantitet 50 mg
Formel vikt 286.33
Renhet >99%
Molekylvikt (g/mol) 286.331