Läs mer
Levo Mepromazine Maleate, TRC
Produktkod. 30138299
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138299 10 mg
30133574 5 mg
17992955 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138299 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL37575010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7104-38-3
Molekylformel C19 H24 N2 O S . C4 H4 O4
Molekylvikt (g/mol) 444.54
Synonym Levomepromazine Maleate, 10H-Phenothiazine-10-propanamine, 2-methoxy-N,N,β-trimethyl-, (R)-, (Z)-2-butenedioate (1:1) (ZCI), Maleic acid, compd. with l-10-(3-dimethylamino-2-methylpropyl)-2-methoxyphenothiazine (6CI), (R)-3-(2-Methoxy-10H-phenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine maleate, Hirnamin, Levomepromazine hydrogen maleate, Levomepromazine maleate, Levopromazine hydrogen maleate, Methotrimeprazine maleate, Milezin, Minozinan, Neruocil, Neuractil, Neurocil, Neurocil (tablet), RP 7044 hydrogen maleate, Sofmin, Veractil
IUPAC-namn (Z)-but-2-enedioic acid;(2R)-3-(2-methoxyphenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine
LEDER COc1ccc2Sc3ccccc3N(C[C@H](C)CN(C)C)c2c1.OC(=O)\C=C/C(=O)O
Specifikationer
Kemiskt namn eller material Levomepromazine Maleate
Kvantitet 10 mg
InChI formel InChI=1S/C19H24N2OS.C4H4O4/c1-14(12-20(2)3)13-21-16-7-5-6-8-18(16)23-19-10-9-15(22-4)11-17(19)21;5-3(6)1-2-4(7)8/h5-11,14H,12-13H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-;/m1./s1
Formel vikt 444.1719