Läs mer
Levocetirizine Ethyl Ester, TRC
Produktkod. 30468192
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30468192 250 mg
1 options
Produktkod. 30468192 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL379260250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Levocetirizine Ethyl Ester
CAS 1116338-10-3
Molekylformel C23 H29 Cl N2 O3
Kvantitet 250 mg
Synonym 2-[2-[4-[(R)-(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]acetic acid ethyl ester
InChI formel InChI=1S/C23H29ClN2O3/c1-2-29-22(27)18-28-17-16-25-12-14-26(15-13-25)23(19-6-4-3-5-7-19)20-8-10-21(24)11-9-20/h3-11,23H,2,12-18H2,1H3/t23-/m1/s1
LEDER CCOC(=O)COCCN1CCN(CC1)[C@H](c2ccccc2)c3ccc(Cl)cc3
IUPAC-namn ethyl 2-[2-[4-[(R)-(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetate
Molekylvikt (g/mol) 416.94
Formel vikt 416.1867