Läs mer
Levodopa Isopropyl Ester, Mikromol™
Produktkod. 30050794
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050794 100 mg
30050495 25 mg
30462146 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30050794 Leverantör Mikromol Leverantörsnummer MM0262.20
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Levodopa Isopropyl Ester
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-DOPA Isopropyl Ester
CAS 110301-07-0
Analyt- eller komponentnamn L-DOPA Isopropyl Ester
Frakt skick Room Temperature
Molekylformel C12 H17 N O4
Certifikat och efterlevnad ISO 17025
Synonym L-Tyrosine, 3-hydroxy-, 1-methylethyl ester, L-DOPA isopropyl ester
InChI formel InChI=1S/C12H17NO4/c1-7(2)17-12(16)9(13)5-8-3-4-10(14)11(15)6-8/h3-4,6-7,9,14-15H,5,13H2,1-2H3/t9-/m0/s1
LEDER CC(C)OC(=O)[C@@H](N)Cc1ccc(O)c(O)c1
IUPAC-namn propan-2-yl (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoate
Molekylvikt (g/mol) 239.27
Kvantitet 100 mg
Formel vikt 239.1158 g/mol