Läs mer
Levofloxacin Ethyl Ester, TRC
Produktkod. 30180402
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180402 1 g
30180159 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180402 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3600201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 177472-30-9
Molekylformel C20 H24 F N3 O4
Molekylvikt (g/mol) 389.42
Synonym (3S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid ethyl ester, 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 9-fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-, ethyl ester, (S)-
IUPAC-namn (3S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid ethyl ester
LEDER CCOC(=O)C1=CN2[C@@H](C)COc3c(N4CCN(C)CC4)c(F)cc(C1=O)c23
Specifikationer
Kemiskt namn eller material Levofloxacin Ethyl Ester
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C20H24FN3O4/c1-4-27-20(26)14-10-24-12(2)11-28-19-16(24)13(18(14)25)9-15(21)17(19)23-7-5-22(3)6-8-23/h9-10,12H,4-8,11H2,1-3H3/t12-/m0/s1
Formel vikt 389.1751
Procent renhet >95%