Läs mer
Levofloxacin Hydrochloride, TRC
Produktkod. 30472345
Change view
Kvantitet:
1 g
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30472345 2.5 g 2.5g
30479454 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30472345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3602582.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Levofloxacin Hydrochloride
CAS 177325-13-2
Molekylformel C18H20FN3O4 . (HCl)
Kvantitet 2.5 g
Synonym Weilisu, (S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid Monohydrochloride, (3S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid Monohydrochloride
InChI formel InChI=1S/C18H20FN3O4.ClH/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21;/h7-8,10H,3-6,9H2,1-2H3,(H,24,25);1H/t10-;/m0./s1
LEDER Cl.C[C@H]1COc2c(N3CCN(C)CC3)c(F)cc4C(=O)C(=CN1c24)C(=O)O
Molekylvikt (g/mol) 397.8
Formel vikt 397.1205