Läs mer
Levomedetomidine, TRC
Produktkod. 30136900
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136900 10 mg
30012641 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136900 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL37570010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 119717-21-4
Molekylformel C13 H16 N2
Molekylvikt (g/mol) 200.28
IUPAC-namn 4-[(1R)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole
LEDER C[C@@H](c1c[nH]cn1)c2cccc(C)c2C
Specifikationer
Kemiskt namn eller material Levomedetomidine
Kvantitet 10 mg
InChI formel InChI=1S/C13H16N2/c1-9-5-4-6-12(10(9)2)11(3)13-7-14-8-15-13/h4-8,11H,1-3H3,(H,14,15)/t11-/m1/s1
Formel vikt 200.1313