Läs mer
Levomefolate Calcium, TRC
Produktkod. 30010996
Change view
Kvantitet:
25 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010996 1 g
30005466 250 mg
17958884 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010996 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3757201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 151533-22-1
Molekylformel C20 H25 N7 O6 . Ca
Molekylvikt (g/mol) 497.52
Synonym L-Glutamic acid, N-[4-[[[(6S)-2-amino-3,4,5,6,7,8-hexahydro-5-methyl-4-oxo-6-pteridinyl]methyl]amino]benzoyl]-, calcium salt (1:1), L-Glutamic acid, N-[4-[[(2-amino-1,4,5,6,7,8-hexahydro-5-methyl-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-, calcium salt (1:1), (S)-, L-Glutamic acid, N-[4-[[[(6S)-2-amino-1,4,5,6,7,8-hexahydro-5-methyl-4-oxo-6-pteridinyl]methyl]amino]benzoyl]-, calcium salt (1:1) (9CI), (6S)-5-Methyltetrahydrofolate calcium salt, Bodyfolin, Calcium (6S)-5-methyl-tetrahydrofolate, Deplin, L-Methylfolate calcium salt, LMCA, Levomefolate calcium, Metafolin, Nutrifolin, Levomefolic acid calcium salt
IUPAC-namn calcium;(2S)-2-[[4-[[(6S)-2-amino-5-methyl-4-oxo-3,6,7,8-tetrahydropteridin-6-yl]methylamino]benzoyl]amino]pentanedioate
LEDER CN1[C@H](CNC2=C1C(=O)NC(=N2)N)CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(=O)[O-])C(=O)[O-].[Ca+2]
Specifikationer
Kemiskt namn eller material Methyl folate calcium salt
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C20H25N7O6.Ca/c1-27-12(9-23-16-15(27)18(31)26-20(21)25-16)8-22-11-4-2-10(3-5-11)17(30)24-13(19(32)33)6-7-14(28)29;/h2-5,12-13,22H,6-9H2,1H3,(H,24,30)(H,28,29)(H,32,33)(H4,21,23,25,26,31);/q;+2/p-2/t12-,13-;/m0./s1
Formel vikt 497.1336
Procent renhet >95%