Läs mer
Levomenol, TRC
Produktkod. 30136967
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136967 1 g
30132398 5 g
17988884 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136967 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3758001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23089-26-1
Molekylformel C15 H26 O
Molekylvikt (g/mol) 222.37
Synonym Levomenol, (-)-α-Bisabolol, 3-Cyclohexene-1-methanol, α,4-dimethyl-α-(4-methyl-3-penten-1-yl)-, (αS,1S)-, 3-Cyclohexene-1-methanol, α,4-dimethyl-α-(4-methyl-3-pentenyl)-, (αS,1S)- (9CI), 3-Cyclohexene-1-methanol, α,4-dimethyl-α-(4-methyl-3-pentenyl)-, [S-(R*,R*)]-, 5-Hepten-2-ol, 6-methyl-2-(4-methyl-3-cyclohexen-1-yl)-, (-)- (8CI), (αS,1S)-α,4-Dimethyl-α-(4-methyl-3-penten-1-yl)-3-cyclohexene-1-methanol, (-)-(1'S,2S)-α-Bisabolol, (-)-(4S,8S)-α-Bisabolol, Kamillosan, l-α-Bisabolol, α-Bisabolol, (-)-, α-(-)-Bisabolol
IUPAC-namn (2S)-6-methyl-2-[(1S)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
LEDER CC(=CCC[C@](C)(O)[C@H]1CCC(=CC1)C)C
Specifikationer
Kemiskt namn eller material alpha-(-)-Bisabolol
Kvantitet 1 g
InChI formel InChI=1S/C15H26O/c1-12(2)6-5-11-15(4,16)14-9-7-13(3)8-10-14/h6-7,14,16H,5,8-11H2,1-4H3/t14-,15+/m1/s1
Formel vikt 222.1984