Läs mer
Levonorgestrel EP Impurity H, Mikromol™
Produktkod. 18082760
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18082760 100 mg
18093560 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18082760 Leverantör Mikromol Leverantörsnummer MM0121.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Levonorgestrel EP Impurity H
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6beta-Hydroxylevonorgestrel
CAS 55555-97-0
Analyt- eller komponentnamn 6beta-Hydroxylevonorgestrel
Frakt skick Room Temperature
Molekylformel C21 H28 O3
Certifikat och efterlevnad ISO 17025
Synonym 13-Ethyl-6β,17-dihydroxy-18,19-dinor-17α-pregn-4-en-20-yn-3-one (6β-Hydroxylevonorgestrel), Levonorgestrel Imp. H (EP), 13-Ethyl-6beta,17-dihydroxy-18,19-dinor-17alpha-pregn-4-en-20-yn-3-one, Levonorgestrel Imp. H (Pharmeuropa), 6beta-Hydroxylevonorgestrel, Levonorgestrel Impurity H
InChI formel InChI=1S/C21H28O3/c1-3-20-9-7-15-14-6-5-13(22)11-17(14)19(23)12-16(15)18(20)8-10-21(20,24)4-2/h2,11,14-16,18-19,23-24H,3,5-10,12H2,1H3/t14-,15-,16-,18+,19-,20+,21+/m1/s1
LEDER CC[C@]12CC[C@H]3[C@@H](C[C@@H](O)C4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
IUPAC-namn (6R,8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-6,17-dihydroxy-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
Molekylvikt (g/mol) 328.45
Kvantitet 100 mg
Formel vikt 328.2038 g/mol