Läs mer
Levopride, TRC
Produktkod. 17954095
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17954095 100 mg
17942095 1 g
17983265 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17954095 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL376000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23672-07-3
Molekylformel C15 H23 N3 O4 S
Molekylvikt (g/mol) 341.43
Synonym 5-(Aminosulfonyl)-N-[[(2S)-1-ethyl-2-pyrrolidinyl]methyl]-2-methoxybenzamide, Benzamide, 5-(aminosulfonyl)-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxy-, (S)-, o-Anisamide, N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-sulfamoyl-, (-)- (8CI), (-)-Sulpiride, L-Sulpiride, Lesuride, Levobren, Levopraid, Levosulpiride, S-(-)-Sulpiride, S-Sulpiride, l-Sulpiride, levopride, sulpid
IUPAC-namn N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2-methoxy-5-sulfamoylbenzamide
LEDER CCN1CCC[C@H]1CNC(=O)c2cc(ccc2OC)S(=O)(=O)N
Specifikationer
Kemiskt namn eller material Levosulpiride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C15H23N3O4S/c1-3-18-8-4-5-11(18)10-17-15(19)13-9-12(23(16,20)21)6-7-14(13)22-2/h6-7,9,11H,3-5,8,10H2,1-2H3,(H,17,19)(H2,16,20,21)/t11-/m0/s1
Formel vikt 341.1409
Procent renhet >95%