missing translation for 'onlineSavingsMsg'
Läs mer

Lewis A Blood Group Antigen Antibody (SPM522) - IHC-Prediluted, Novus Biologicals™

Produktkod. 18177888 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18177888 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18177888 Leverantör Novus Biologicals Leverantörsnummer NBP248080

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

Lewis A Blood Group Antigen Monoclonal specifically detects Lewis A Blood Group Antigen in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Lewis A Blood Group Antigen
Användningsområden Immunohistochemistry (Paraffin)
Klassificering Monoclonal
Klona SPM522
Konjugera Unconjugated
Utspädning Immunohistochemistry-Paraffin
Formulering 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias A transferase, A3GALNT, A3GALT1, ABO, B transferase, EC=2.4.1.37, EC=2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, Histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT
Gensymboler ABO
Värd art Mouse
Immunogen Mucins isolated from ovarian cyst fluid
Reningsmetod Protein A or G purified
Kvantitet 7 mL
Regulatorisk status RUO
Primär eller sekundär Primary
Testspecificitet Recognizes a carbohydrate determinant of Gal 1-3(Fuc 1-4) GlcNAc which is blood group antigen Lewis A. It is present primarily on epithelial cells such as colon and kidneys. In the tumors and dedifferentiated tissues, decrease of Lewis A antigen was observed. Lewis A (type 1 chain) is expressed in colonic epithelial cells and may be useful for detection of gastrointestinal tumors, pancreatic cancer, and colorectal tumors. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
Målarter Human, Mouse
Innehåll och lagring Store at 4C.
Form Purified
Isotyp IgG1 κ
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.