Läs mer
Lewis A Trisaccharide, TRC
Produktkod. 30480370
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480370 5 mg
30474831 10 mg
30465394 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30480370 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3940005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56570-03-7
Molekylformel C20 H35 N O15
Molekylvikt (g/mol) 529.49
IUPAC-namn N-[(3R,4R,5S,6R)-2-hydroxy-6-(hydroxymethyl)-4-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-5-[(2S,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-3-yl]acetamide
LEDER C[C@@H]1O[C@@H](O[C@@H]2[C@@H](CO)OC(O)[C@H](NC(=O)C)[C@H]2O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material Lewis A Trisaccharide
Kvantitet 5 mg
InChI formel InChI=1S/C20H35NO15/c1-5-10(25)12(27)14(29)19(32-5)35-16-8(4-23)33-18(31)9(21-6(2)24)17(16)36-20-15(30)13(28)11(26)7(3-22)34-20/h5,7-20,22-23,25-31H,3-4H2,1-2H3,(H,21,24)/t5-,7+,8+,9+,10+,11-,12+,13-,14-,15+,16+,17+,18?,19-,20-/m0/s1
Formel vikt 529.2007