Läs mer
Lexacalcitol, MedChemExpress
Beskrivning
Lexacalcitol (KH1060), a vitamin D analog, is a potent regulator of cell growth and immune responses. Lexacalcitol can be used for the research of graft rejection, psoriasis, cancer and auto-immune diseases.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Lexacalcitol |
| CAS | 131875-08-6 |
| Färg | White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C29H48O4 |
| Kvantitet | 1 mg |
| Synonym | KH1060 |
| Löslighetsinformation | DMSO : 100 mg/mL (217.07 mM; Need ultrasonic) |
| LEDER | C[C@]1(CCC/2)[C@](CC[C@@H]1[C@@H](C)OCCCC(CC)(CC)O)([H])C2=C\C=C(C[C@@H](O)C[C@@H]3O)/C3=C |
| Molekylvikt (g/mol) | 460.69 |
| Visa mer |