missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567868
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse and Rat samples.
LGR4 Polyclonal antibody specifically detects LGR4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
LGR4 | |
Polyclonal | |
Unconjugated | |
LGR4 | |
9130225G07; A330106J01Rik; A930009A08Rik; BNMD17; G protein-coupled receptor 48; G protein-coupled receptor GPR48; GPR48; G-protein coupled receptor 48; leucine rich repeat containing G protein-coupled receptor 4; leucine-rich repeat containing G protein-coupled receptor 4; leucine-rich repeat-containing G protein-coupled receptor 4; leucine-rich repeat-containing G-protein coupled receptor 4; Lgr4 | |
Rabbit | |
affinity chromatography | |
RUO | |
107515, 286994, 55366 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A2ARI4, Q9BXB1, Q9Z2H4 | |
LGR4 | |
Synthesized peptide derived from human LGR4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |