Learn More
Lgr6 Antibody (918726), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB84581H
Product Code. 18610535
  • 5960.00 SEK / 0.10ml
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is not returnable. View return policy
Description
Description
Lgr6 Monoclonal antibody specifically detects Lgr6 in Human samples. It is validated for Flow Cytometry, Blocking Assay, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lgr6
Monoclonal
HRP
PBS
Mouse
0.1 mL
GPCR
59352
Store at 4C in the dark.
Flow Cytometry, Blocking Assay, Immunocytochemistry, CyTOF
918726
Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready
FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631
NS0 mouse myeloma cell line transfected with human Lgr6, Met1-Val967, Accession # Q9HBX8
RUO
Primary
Human
IgG2a