Learn More
LHPP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552658
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LISLGKGRYY KETSGLMLDV GPYMKALEYA CGIKAEVVGK PSPEFFKSAL QAIGVEAHQA VMIGDDIVGD VGGAQRCGMR ALQVRTGKFR PSDEHHPEVK ADGYVD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.
LHPP Polyclonal antibody specifically detects LHPP in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
LHPP | |
Polyclonal | |
Unconjugated | |
Lhpp | |
2310007H09Rik; HDHD2B; hLHPP; LHPP; Phospholysine phosphohistidine inorganic pyrophosphate phosphatase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
361663, 64077, 76429 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5I0D5, Q9D7I5, Q9H008 | |
Lhpp | |
Recombinant protein corresponding to Human LHPP | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only