missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ LHR Polyclonal Antibody

Beskrivning
Immunogen sequence: SVTLKLYGNG FEEVQSHAFN GTTLTSLELK ENVHLEKMHN GAFRGATGPK TLDISSTKLQ ALPSYGLESI QR Antigen sequence identity: Mouse 83%; Rat 64%.
LHCGR encodes the receptor for both luteinizing hormone and choriogonadotropin. It is family 1 of G-protein coupled receptors, FSH/LSH/TSH superfamily, and its basic function is acting as luteinizing hormone/choriogonadotropin receptor (gonadotropin receptor). Regulating the development and function of Leyding cells in males. It also plays a role in male secondary sexual characterdevelopment. Mutations in this gonadotropinreceptor results in disorders of male secondary sexual characteristic development, such as familial male precocious puperty and male pseudohermaphtoditism.
Specifikationer
Specifikationer
| Antigen | LHR |
| Användningsområden | Immunocytochemistry |
| Klassificering | Polyclonal |
| Koncentration | 0.3 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gen | LHCGR |
| Genaccessionsnr. | P22888 |
| Gene Alias | Gpcr19-rs1; HCG receptor; HHG; hypergonadotropic hypogonadism; LCGR; LGR2; LH receptor; LH/CGR; LH/CG-R; LHCGR; LHR; LH-R; LHRHR; LSHR; LSH-R; Luteinizing hormone receptor; luteinizing hormone/choriogonadotropin receptor; Lutropin receptor; lutropin/choriogonadotropin receptor; lutropin-choriogonadotropic hormone receptor; ULG5 |
| Gensymboler | LHCGR |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?