missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11577-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
LIAS belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis.The protein encoded by this gene belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.The protein encoded by this gene belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.Specifications
LIAS | |
Polyclonal | |
Unconjugated | |
LIAS | |
HUSSY 01, LAS, LIAS, Lip syn, LIP1, Lipoate synthase, Lipoic acid synthase, lipoic acid synthetase, Lipoyl synthase, mitochondrial, LS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11019, 305348, 79464 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43766, Q5XIH4, Q99M04 | |
Lias | |
LIAS Fusion Protein Ag2162 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |