missing translation for 'onlineSavingsMsg'
Learn More

LIAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11577-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828753

  • 4721.67 SEK / 150µL
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

LIAS belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis.The protein encoded by this gene belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.The protein encoded by this gene belongs to the biotin and lipoic acid synthetases family. It localizes in mitochondrion and plays an important role in alpha-(+)-lipoic acid synthesis. It may also function in the sulfur insertion chemistry in lipoate biosynthesis. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications

Specifications

LIAS
Polyclonal
Unconjugated
LIAS
HUSSY 01, LAS, LIAS, Lip syn, LIP1, Lipoate synthase, Lipoic acid synthase, lipoic acid synthetase, Lipoyl synthase, mitochondrial, LS
Rabbit
Antigen Affinity Chromatography
RUO
11019, 305348, 79464
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43766, Q5XIH4, Q99M04
Lias
LIAS Fusion Protein Ag2162
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers