Learn More
Buerkle™ HDPE Lid for Round Tub
Buerkle™ HDPE Lid for Round Tub is suitable for use with Buerkle™ Tubs.
Brand: Buerkle™ 3482-0401
203.74 SEK valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.36000kg
Description
- Accessory for round tubs
- Chemical resistance
- Suitable for food
- Made of HDPE
Specifications
Lid | |
370 mm | |
1 Ea. |
Polyethylene | |
For Round Tubs | |
-30°C to 70°C |