Läs mer
Lidamidine Hydrochloride, TRC
Produktkod. 30477083
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30477083 100 mg
1 options
Produktkod. 30477083 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL397790100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lidamidine Hydrochloride
CAS 65009-35-0
Molekylformel C11 H16 N4 O . Cl H
Kvantitet 100 mg
Synonym N-(2,6-Dimethylphenyl)-N'-[imino(methylamino)methyl]urea Hydrochloride (1:1), Urea, N-(2,6-dimethylphenyl)-N'-[imino(methylamino)methyl]-, monohydrochloride (9CI), Lidamidine Hydrochloride, Lidarral, Smodin, WHJR 1142A, WHR 1142A
InChI formel InChI=1S/C11H16N4O.ClH/c1-7-5-4-6-8(2)9(7)14-11(16)15-10(12)13-3;/h4-6H,1-3H3,(H4,12,13,14,15,16);1H
LEDER Cl.CNC(=N)NC(=O)Nc1c(C)cccc1C
IUPAC-namn 1-(2,6-dimethylphenyl)-3-(N-methylcarbamimidoyl)urea;hydrochloride
Molekylvikt (g/mol) 256.73
Formel vikt 256.1091