Läs mer
Lidocaine N-Oxide, TRC
Produktkod. 18021410
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18021410 100 mg
18030030 10 mg
18073610 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18021410 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL397815100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2903-45-9
Molekylformel C14 H22 N2 O2
Molekylvikt (g/mol) 250.34
Synonym 2-(Diethylazinoyl)-N-(2,6-dimethylphenyl)acetamide, Lidocaine Hydrochloride Imp. B (EP), Lidocaine N2-Oxide, Lidocaine N-Oxide, 2-(Diethyloxidoamino)-N-(2,6-dimethylphenyl)acetamide
IUPAC-namn 2-(2,6-dimethylanilino)-N,N-diethyl-2-oxoethanamine oxide
LEDER CC[N+]([O-])(CC)CC(=O)Nc1c(C)cccc1C
Specifikationer
Kemiskt namn eller material Lidocaine N-Oxide (2-(Diethyloxidoamino)-N-(2,6-dimethylphenyl)acetamide)
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C14H22N2O2/c1-5-16(18,6-2)10-13(17)15-14-11(3)8-7-9-12(14)4/h7-9H,5-6,10H2,1-4H3,(H,15,17)
Formel vikt 250.1681
Procent renhet >95%