missing translation for 'onlineSavingsMsg'
Läs mer

Lidocaine N-Oxide, TRC

Produktkod. 18073610
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18073610 25 mg
18021410 100 mg
18030030 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18073610 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL39781525MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2903-45-9
Molekylformel C14 H22 N2 O2
Molekylvikt (g/mol) 250.34
Synonym 2-(Diethylazinoyl)-N-(2,6-dimethylphenyl)acetamide, Lidocaine Hydrochloride Imp. B (EP), Lidocaine N2-Oxide, Lidocaine N-Oxide, 2-(Diethyloxidoamino)-N-(2,6-dimethylphenyl)acetamide
IUPAC-namn 2-(2,6-dimethylanilino)-N,N-diethyl-2-oxoethanamine oxide
LEDER CC[N+]([O-])(CC)CC(=O)Nc1c(C)cccc1C

Specifications

Kemiskt namn eller material Lidocaine N-Oxide (2-(Diethyloxidoamino)-N-(2,6-dimethylphenyl)acetamide)
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C14H22N2O2/c1-5-16(18,6-2)10-13(17)15-14-11(3)8-7-9-12(14)4/h7-9H,5-6,10H2,1-4H3,(H,15,17)
Formel vikt 250.1681
Procent renhet >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.