Learn More
Invitrogen™ E-Gel™ Imager iBase™ Light Diffuser
Improves quality of gel images from the E-Gel™ iBase™ Power Base when placed on the E-Gel Imager Blue-Light Base or E-Gel Safe Imager™
Brand: Invitrogen™ 4473062
Additional Details : Weight : 0.00450kg
Includes: 1 E-Gel™ Imager iBase Light Diffuser
Description
Imaging gels without using the Light Diffuser may result in image artifacts that interfere with result analysis. This is a replacement part for the Light Diffuser included in the E-Gel Imager System.
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
1 E-Gel™ Imager iBase Light Diffuser Store at room temperature |
|
E-Gel | |
Room Temperature |
E-Gel Imager | |
Imager Light Diffuser |
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.