Learn More
Eppendorf™ Lower Part for Eppendorf Research™ plus Pipette
For use with Eppendorf Research plus Pipettes. Designed for maximum safety, robustness and ergonomics
Brand:  Eppendorf™ 3120615008
Additional Details : Weight : 0.03100kg
 View more versions of this product
Product Code. 10014924
  • 516.00 SEK / Each
Estimated Shipment
14-08-2018
Add to basket
Description
Description
  • Eppendorf Research plus Pipettes
  • Ultralight pipette
  • Developed according to the strict criteria of the Eppendorf PhysioCare Concept
  • Eppendorf PerfectPiston™: Ultralight and highly resistant piston system
  • Positions and shapes of the control buttons/displays have been optimized for stress-free work procedures and intuitive operation
  • Spring-loaded tip cone provides you with an optimal pipette fit with minimal insertion force (not available for 5mL and 10mL pipettes)
  • Adjustment option allows you to perfectly adapt the pipette for specific liquids or other external conditions
  • New volume setting can switch from maximum to minimum volume in just a few turns
  • 4-digit volume display with magnifying display makes quick and accurate volume settings easier than ever
  • Quick lock makes the lower part particularly easy to remove and service (not available for 5mL and 10mL pipettes)
Specifications
Specifications
Eppendorf Research plus Pipette
Eppendorf Research plus Pipette
Single channel
2 to 20μL
1Each
Light Grey
Lower Part for Eppendorf Research plus Pipettes
Videos