Läs mer
Novus Biologicals™ Lightning-Link (R) Rapid Alexa Fluor 594 Antibody Labeling Kit
Handla allt Bio Techne Produkter
Kvantitet:
1 mg
100 μg
30 μg
300 μg
Beskrivning
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
- Lightning-Link™ Rapid mix
- 1 vial of LL Rapid Quencher reagent
- 1 vial of LL Rapid Modifier reagent
Specifikationer
Specifikationer
| Kvantitet | 1 mg |
| Produkttyp | Antibody Labeling Kit |
| Innehåll och lagring | -20°C |