Läs mer
Ligustroside, TRC
Produktkod. 30483009
Change view
Kvantitet:
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30483009 10 mg -
30469298 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30483009 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL39796010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ligustroside
CAS 35897-92-8
Anteckningar om renhetsgrad HPLC
Molekylformel C25H32O12
Kvantitet 10 mg
Synonym (2S,3E,4S)-3-Ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-2H-pyran-4-acetic Acid 2-(4-Hydroxyphenyl)ethyl Ester, [2S-(2α,3E,4β)]-3-Ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-2H-pyran-4-acetic Acid 2-(4-Hydroxyphenyl)ethyl Ester, Ligstroside
InChI formel InChI=1S/C25H32O12/c1-3-15-16(10-19(28)34-9-8-13-4-6-14(27)7-5-13)17(23(32)33-2)12-35-24(15)37-25-22(31)21(30)20(29)18(11-26)36-25/h3-7,12,16,18,20-22,24-27,29-31H,8-11H2,1-2H3/b15-3-/t16-,18?,20+,21-,22?,24-,25-/m0/s1
LEDER COC(=O)C1=CO[C@@H](O[C@@H]2OC(CO)[C@@H](O)[C@H](O)C2O)\C(=C/C)\[C@@H]1CC(=O)OCCc3ccc(O)cc3
IUPAC-namn methyl (4S,5Z,6S)-5-ethylidene-4-[2-[2-(4-hydroxyphenyl)ethoxy]-2-oxo-ethyl]-6-[(2S,4S,5S)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl]oxy-4H-pyran-3-carboxylate
Molekylvikt (g/mol) 524.51
Formel vikt 524.1894