Läs mer
Limaprost, TRC
Produktkod. 30477501
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477501 1 mg
30476592 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30477501 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4615001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74397-12-9
Molekylformel C22 H36 O5
Molekylvikt (g/mol) 380.52
Synonym 2-Heptenoic acid, 7-[(1R,2R,3R)-3-hydroxy-2-[(1E,3S,5S)-3-hydroxy-5-methyl-1-nonen-1-yl]-5-oxocyclopentyl]-, (2E)- (ACI), (2E)-7-[(1R,2R,3R)-3-Hydroxy-2-[(1E,3S,5S)-3-hydroxy-5-methyl-1-nonen-1-yl]-5-oxocyclopentyl]-2-heptenoic acid (ACI), 2-Heptenoic acid, 7-[(1R,2R,3R)-3-hydroxy-2-[(1E,3S,5S)-3-hydroxy-5-methyl-1-nonenyl]-5-oxocyclopentyl]-, (2E)- (9CI), Prosta-2,13-dien-1-oic acid, 11,15-dihydroxy-17,20-dimethyl-9-oxo-, (2E,11α,13E,15S,17S)- (ZCI), 17S,20-Dimethyl-trans-δ2-PGE1, Limaprost, ONO 1206, OP 1206
IUPAC-namn (E)-7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S,5S)-3-hydroxy-5-methylnon-1-enyl]-5-oxocyclopentyl]hept-2-enoic acid
LEDER CCCC[C@H](C)C[C@H](O)\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCC\C=C\C(=O)O
Specifikationer
Kemiskt namn eller material Limaprost
Kvantitet 1 mg
InChI formel InChI=1S/C22H36O5/c1-3-4-9-16(2)14-17(23)12-13-19-18(20(24)15-21(19)25)10-7-5-6-8-11-22(26)27/h8,11-13,16-19,21,23,25H,3-7,9-10,14-15H2,1-2H3,(H,26,27)/b11-8+,13-12+/t16-,17+,18+,19+,21+/m0/s1
Formel vikt 380.2563