missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN28 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | LIN28 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838993
|
Proteintech
11724-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16828993
|
Proteintech
11724-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is a rabbit polyclonal antibody raised against full length LIN28 of human origin. The calculated molecular weight of LIN28 is 23 kDa, but the modified LIN28 is about 28 kDa.
Lin28 is a transcription factor that was first identified through its key role in the pathway of developmental timing in C. elegans. The role of Lin28 in development suggested that it might be useful in the creation of stem cells that might be beneficial in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing Lin28 in addition to the transcription factors POU5F1, Sox2, and NANOG in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing Lin28, Sox2, POU5F1 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.Specifications
LIN28 | |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q8K3Y3, Q9H9Z2 | |
500562, 79727, 83557 | |
LIN28 Fusion Protein Ag2312 | |
Primary | |
-20°C | |
LIN28A |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CSDD1, LIN 28, lin 28 homolog (C. elegans), Lin 28A, LIN28, Lin28A, Protein lin 28 homolog A, ZCCHC1 | |
Lin28a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |