missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN28B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24017-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.Specifications
LIN28B | |
Polyclonal | |
Unconjugated | |
LIN28B | |
CSDD2, lin 28 homolog B (C. elegans), Lin 28B, LIN28B, Protein lin 28 homolog B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
380669, 389421 | |
-20°C | |
Liquid |
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q45KJ6, Q6ZN17 | |
Lin28b | |
LIN28B Fusion Protein Ag21241 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |