Learn More
LIN9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556412
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LNKDLNKVLH KVQQYCYELA PDQGLQPADQ PTDMRRRCEE EAQEIVRHAN SSTGQPCVEN ENLTDLISRL TAILLQIKCL A Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
LIN9 Polyclonal antibody specifically detects LIN9 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
LIN9 | |
Polyclonal | |
Unconjugated | |
LIN9 | |
2700022J23Rik; BARA; BARPsv; Beta subunit-associated regulator of apoptosis; HuLin-9; LIN9; Lin-9; lin-9 DREAM MuvB core complex component; lin-9 homolog; lin-9 homolog (C. elegans); mLin-9; pRB-associated protein; protein lin-9 homolog; rb related pathway actor; TGS; TGS1; TGS2; TUDOR gene similar 1 protein; TUDOR gene similar protein; Type I interferon receptor beta chain-associated protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
286826 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TKA1 | |
LIN9 | |
Recombinant protein corresponding to Human LIN9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only